Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079835.2 | NP_524574.1 | 1 - 2 | 248 | 100 | |
GenBank mRNA | AK136521.1 | - | 1 - 2 | 250 | 100 | |
BC141103.1 | - | 1 - 2 | 138 | 9055715 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |