Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145827.3 | NP_665826.1 | 6 - 7 | 2716 | 87 | |
GenBank mRNA | AF486632.1 | - | 7 - 8 | 2612 | 87 | |
AY337285.1 | - | 5 - 6 | 2502 | 87 | ||
AY355340.1 | - | 5 - 6 | 2490 | 87 | ||
AY495376.1 | - | 6 - 7 | 2716 | 87 | ||
AY495377.1 | - | 7 - 8 | 2612 | 87 | ||
BC116174.1 | - | 6 - 7 | 2676 | 40048724 | 87 | |
KF032621.1 | - | 5 - 6 | 2492 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |