Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mirlet7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_029727.1 | 1 | 46 | 76 |
Gene Symbol: Lincppara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110483.1 | 2 | 1329 | 76 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA let7c-2 | Mirlet7c-2 |
Mirnlet7c-2, let-7c-2 |
-622.0 | Sense | Chr. 15 - 85706603 - 85706697 on NCBI Build GRCm38 |
Downstream | peroxisome proliferator activated receptor alpha | Ppara |
4933429D07Rik, AW742785, Nr1c1, PPAR-alpha, PPARalpha, Ppar |
20.8kb | Sense | Chr. 15 - 85728192 - 85806851 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |