Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art2a-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033804.1 | 3 - 4 | 854 | 80 | ||
GenBank mRNA | AF016464.1 | - | 2 - 3 | 784 | 80 | |
AK018555.1 | - | 3 - 4 | 855 | 80 | ||
S79913.1 | - | 796 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 35363 |
Gm35363 | -13.8kb | Sense | Chr. 7 - 101521064 - 101538664 on NCBI Build GRCm38 | |
Downstream | ADP-ribosyltransferase 2b | Art2b |
ART2.2, ARTC2, Art, Ly92b, Rt-6, Rt6, Rt6-2 |
20.5kb | Sense | Chr. 7 - 101575729 - 101585583 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |