Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
TC0700000806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_001205073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_025368.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006668.mm.1 |
NM_025368.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
TC0700000806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_001205073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_025368.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006669.mm.1 |
NM_025368.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
TC0700000806.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205070.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205070.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205071.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205071.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205072.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205072.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205073.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_001205073.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_025368.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006670.mm.1 |
NM_025368.4 |
Refseq |