Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430562O15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015515.2 | 1 - 2 | 987 | 145 | ||
GenBank mRNA | AK031668.1 | - | 1 - 2 | 987 | 145 | |
AK032482.1 | - | 1038 | 145 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitochondrial ribosomal protein S27 | Mrps27 | 2610028H14Rik | Anti-sense | Chr. 13 - 99344767 - 99415562 on NCBI Build GRCm38 | |
Downstream | microtubule-associated protein 1B | Map1b |
A230055D22, AI843217, LC1, MAP5, Mtap-5, Mtap1b, Mtap5 |
8.6kb | Sense | Chr. 13 - 99421464 - 99516602 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |