Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199212.1 | NP_001186141.1 | 1 - 2 | 137 | 68 | |
GenBank mRNA | AB512670.1 | - | 1 - 2 | 112 | 68 | |
AB512671.1 | - | 1 - 2 | 137 | 68 | ||
AB512672.1 | - | 1 - 2 | 89 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |