Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199212.1 | NP_001186141.1 | 5 - 6 | 632 | 103 | |
GenBank mRNA | AB512670.1 | - | 5 - 6 | 607 | 103 | |
AB512671.1 | - | 5 - 6 | 632 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |