Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oprl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045542.1 | 3 - 4 | 490 | 100 | ||
GenBank mRNA | AK044178.1 | - | 3 - 4 | 425 | 100 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | LKAAEAR motif containing 1 (IKAAEAR murine motif) | Lkaaear1 |
4930526D03Rik, Gm692 |
-18.0kb | Anti-sense | Chr. 2 - 181696793 - 181698468 on NCBI Build GRCm38 |
Downstream | myelin transcription factor 1 | Myt1 |
Nzf2, Nzf2a, Nzf2b, Nztf2 |
44.6kb | Sense | Chr. 2 - 181763332 - 181827781 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |