Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300000528.mm.1 |
NR_045575.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000528.mm.1 |
NR_045575.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000528.mm.1 |
TC0300000124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NM_001253390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NM_001253390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NM_001253391.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NM_001253391.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NM_026182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NM_026182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NR_045575.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
NR_045575.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000525.mm.1 |
TC0300000124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NM_001253390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NM_001253390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NM_001253391.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NM_001253391.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NM_026182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NM_026182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NR_045575.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
NR_045575.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000527.mm.1 |
TC0300000124.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300000519.mm.1 |
NM_026182.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000519.mm.1 |
NM_026182.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000519.mm.1 |
NR_045575.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000519.mm.1 |
NR_045575.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300000519.mm.1 |
TC0300000124.mm.1 |
MTA TC |