Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Milr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073165.1 | 4 - 5 | 593 | 73 | ||
GenBank mRNA | AK137500.1 | - | 4 - 5 | 552 | 73 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
polymerase (DNA directed), gamma 2, accessory subunit |
Polg2 | Anti-sense | Chr. 11 - 106754821 - 106780010 on NCBI Build GRCm38 | ||
Downstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 5 | Ddx5 |
2600009A06Rik, G17P1, HUMP68, Hlr1, p68 |
19.1kb | Anti-sense | Chr. 11 - 106780355 - 106789268 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |