Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iba57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173785.6 | NP_776146.1 | 2 - 3 | 740 | 116 | |
GenBank mRNA | AK038625.1 | - | 2 - 3 | 735 | 116 | |
AK133203.1 | - | 2 - 3 | 738 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons |