Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102382.1 | 7 - 8 | 670 | 86 | ||
GenBank mRNA | AK020981.1 | - | 3 - 4 | 275 | 86 | |
CN455644.1 | - | 6 - 7 | 456 | 86 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 5730522E02 gene | 5730522E02Rik | -223.9kb | Anti-sense | Chr. 11 - 25616846 - 26210576 on NCBI Build GRCm38 | |
Downstream | vaccinia related kinase 2 | Vrk2 |
2810003O05Rik, AI447698 |
3.0kb | Anti-sense | Chr. 11 - 26471401 - 26594014 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |