Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptgs2os2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110420.1 | 1 - 2 | 137 | 107 | ||
GenBank mRNA | AK144630.1 | - | 1 - 2 | 138 | 107 | |
JX682706.1 | - | 1 - 2 | 137 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prostaglandin-endoperoxide synthase 2 | Ptgs2 |
COX2, Cox-2, PGHS-2, PHS-2, Pghs2, TIS10 |
-51.0kb | Anti-sense | Chr. 1 - 150100124 - 150108017 on NCBI Build GRCm38 |
Downstream | phosducin | Pdc |
Rpr-1, Rpr1 |
154.5kb | Anti-sense | Chr. 1 - 150319417 - 150333906 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |