Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195316.1 | NP_001182245.1 | 3 - 4 | 378 | 89 | |
XM_012100410.2 | XP_011955800.1 | 4 - 5 | 511 | 89 | ||
GenBank mRNA | EU370536.1 | - | 3 - 4 | 378 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |