Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246212.1 | NP_001233141.1 | 4 - 5 | 588 | 94 | |
XM_027964811.1 | XP_027820612.1 | 4 - 5 | 545 | 94 | ||
GenBank mRNA | FJ915125.1 | - | 4 - 5 | 588 | 94 |