Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019557.4 | XP_004019606.2 | 3 - 4 | 1088 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |