Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162545.1 | NP_001156017.1 | 2 - 3 | 262 | 79 | |
XM_012158906.3 | XP_012014296.1 | 3 - 4 | 576 | 79 | ||
GenBank mRNA | FJ969413.1 | - | 2 - 3 | 262 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |