Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015100.4 | XP_004015149.2 | 3 - 4 | 524 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |