Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015277.4 | XP_004015326.3 | 1 - 2 | 201 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |