Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969768.1 | XP_027825569.1 | 2 - 3 | 232 | 74 | |
XM_027969769.1 | XP_027825570.1 | 3 - 4 | 475 | 74 | ||
XM_027969770.1 | XP_027825571.1 | 3 - 4 | 346 | 74 |