Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959605.1 | XP_027815406.1 | 3 - 4 | 382 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |