Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010055.4 | XP_004010104.3 | 2 - 3 | 1410 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |