Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267884.2 | NP_001254813.1 | 3 - 4 | 452 | 89 | |
XM_012110711.2 | XP_011966101.1 | 4 - 5 | 466 | 89 | ||
GenBank mRNA | JQ780322.1 | - | 3 - 4 | 456 | 89 |