Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009224.1 | NP_001009224.1 | 297 | 66 | ||
GenBank mRNA | AF152416.1 | - | 297 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |