Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164492.1 | NP_001157964.1 | 4 - 5 | 421 | 85 | |
XM_027977368.1 | XP_027833169.1 | 4 - 5 | 2129 | 85 | ||
GenBank mRNA | EU627104.1 | - | 4 - 5 | 421 | 85 |