Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308589.1 | NP_001295518.1 | 1 - 2 | 578 | 119 | |
GenBank mRNA | KF873613.1 | - | 1 - 2 | 578 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |