Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KISS1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318077.1 | NP_001305006.1 | 1 - 2 | 243 | 66 | |
GenBank mRNA | EU272412.1 | - | 1 - 2 | 27 | 66 | |
JQ812137.1 | - | 1 - 2 | 243 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Ovis aries |