Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040288.2 | NP_001035378.1 | 1 - 2 | 95 | 83 | |
GenBank mRNA | DQ228866.1 | - | 1 - 2 | 99 | 83 | |
MG669627.1 | - | 1 - 2 | 77 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |