Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163051.1 | NP_001156523.1 | 2 - 3 | 360 | 103 | |
XM_012097268.3 | XP_011952658.2 | 3 - 4 | 859 | 103 | ||
XM_012097269.3 | XP_011952659.2 | 3 - 4 | 420 | 103 | ||
GenBank mRNA | FJ958366.1 | - | 2 - 3 | 360 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |