Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIRAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127282.1 | NP_001120754.1 | 1 | 325 | 89 | |
XM_012176689.3 | XP_012032079.1 | 2 | 622 | 89 | ||
GenBank mRNA | EU626201.1 | - | 1 | 325 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |