Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127285.1 | NP_001120757.1 | 3 - 4 | 347 | 88 | |
XM_012097249.3 | XP_011952639.1 | 6 - 7 | 7334 | 88 | ||
XM_012097252.3 | XP_011952642.1 | 5 - 6 | 7298 | 88 | ||
XM_012097256.3 | XP_011952646.1 | 6 - 7 | 7086 | 88 | ||
XM_012097257.3 | XP_011952647.1 | 6 - 7 | 7446 | 88 | ||
XM_012097260.3 | XP_011952650.1 | 8 - 9 | 3911 | 88 | ||
XM_027980221.1 | XP_027836022.1 | 6 - 7 | 5629 | 88 | ||
XM_027980222.1 | XP_027836023.1 | 6 - 7 | 4472 | 88 | ||
XM_027980223.1 | XP_027836024.1 | 7 - 8 | 3995 | 88 | ||
GenBank mRNA | EU626204.1 | - | 3 - 4 | 347 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |