Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142891.1 | NP_001136363.1 | 4 - 5 | 329 | 95 | |
GenBank mRNA | DQ377365.1 | - | 4 - 5 | 329 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |