Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078659.1 | NP_001072127.1 | 4 - 5 | 372 | 106 | |
GenBank mRNA | AY995197.1 | - | 4 - 5 | 372 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |