Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009469.1 | NP_001009469.1 | 9 - 10 | 1197 | 99 | |
XM_027973955.1 | XP_027829756.1 | 10 - 11 | 1311 | 99 | ||
XM_027973956.1 | XP_027829757.1 | 10 - 11 | 1396 | 99 | ||
XM_027973957.1 | XP_027829758.1 | 10 - 11 | 1237 | 99 | ||
GenBank mRNA | X80024.1 | - | 1197 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |