Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001280691.1 | NP_001267620.1 | 2 - 3 | 768 | 71 | |
GenBank mRNA | KC857456.1 | - | 2 - 3 | 768 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |