Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306125.1 | NP_001293054.1 | 1 - 2 | 329 | 72 | |
XM_027965735.1 | XP_027821536.1 | 4 - 5 | 950 | 72 | ||
XM_027965736.1 | XP_027821537.1 | 2 - 3 | 1144 | 72 | ||
GenBank mRNA | KF019682.1 | - | 1 - 2 | 329 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |