Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009809.1 | NP_001009809.1 | 8 - 9 | 1064 | 74 | |
XM_012111144.2 | XP_011966534.1 | 9 - 10 | 1394 | 74 | ||
GenBank mRNA | Z29372.1 | - | 1064 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |