Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127275.1 | NP_001120747.1 | 1 - 2 | 71 | 57 | |
XM_027957545.1 | XP_027813346.1 | 1 - 2 | 386 | 57 | ||
GenBank mRNA | EU626218.1 | - | 1 - 2 | 71 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |