Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190219.3 | XP_012045609.1 | 4 - 5 | 437 | 94 | |
GenBank mRNA | DQ449954.1 | - | 1 - 2 | 81 | 94 |