Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102002.2 | XP_014957488.2 | 4 - 5 | 4154 | 111 | |
XM_027957236.1 | XP_027813037.1 | 4 - 5 | 4312 | 111 | ||
XM_027957237.1 | XP_027813038.1 | 4 - 5 | 4311 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |