Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963490.1 | XP_027819291.1 | 9 - 10 | 1036 | 67 |