Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013811.3 | XP_004013860.1 | 3 - 4 | 1260 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |