Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015098.4 | XP_004015147.1 | 2 - 3 | 357 | 72 | |
XM_015100516.2 | XP_014956002.1 | 2 - 3 | 361 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |