Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003192.4 | XP_004003241.1 | 2 - 3 | 1655 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |