Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002010.4 | XP_004002059.1 | 5 - 6 | 1017 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |