Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016513.4 | XP_004016562.2 | 4 - 5 | 462 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |