Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979655.1 | XP_027835456.1 | 1 - 2 | 1164 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |