Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027964045.1 | XP_027819846.1 | 3 - 4 | 412 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |