Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004013832.4 | XP_004013881.1 | 6 - 7 | 1550 | 104 | |
XM_015099338.2 | XP_014954824.1 | 6 - 7 | 1144 | 104 | ||
XM_015099339.2 | XP_014954825.1 | 6 - 7 | 2500 | 104 | ||
XM_027975910.1 | XP_027831711.1 | 5 - 6 | 635 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |